Order Entry
France
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Alkylphenol Target Analyte Mix Solution For Din En Iso 18857-2
Catalog # 33623-10ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Alkylphenol Target Analyte Mix Solution For Din En Iso 18857-2
Catalog # 33623-10ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Standards:
    Analytical standard
  • Description:
    Alkylphenol Target Analyte Mix solution for DIN EN ISO 18857-2
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H319 - H336
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Storage temperature:
    −20 °C
  • Concentration:
    In acetone
  • Flash point:
    –17,0 °C - closed cup - Solvent

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338