Order Entry
France
ContactUsLinkComponent
Sodium-1-decanesulphonate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Sodium-1-decanesulphonate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Catalog # 85792.150
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium-1-decanesulphonate ≥99.0%, HiPerSolv CHROMANORM® for HPLC
Catalog # 85792.150
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (calculated on dried substance):
    ≥ 99,00 %
  • Appearance:
    White to cream powder
  • IR Spectrum:
    Passes test
  • Appearance of solution (5 %):
    Clear and colourless
  • Water:
    ≤ 1,0 %
  • Absorbance (210 nm) (0.2 mol/l):
    ≤ 0,05
  • Absorbance (220 nm) (0.2 mol/l):
    ≤ 0,03
  • Absorbance (230 nm) (0.2 mol/l):
    ≤ 0,02
  • Absorbance (260 nm) (0.2 mol/l):
    ≤ 0,02

Specification Test Results

About this item

 :
 :244.33 g/mol
Storage Temperature:Ambient
CAS Number:13419-61-9
Formula:C₁₀H₂₁NaO₃S
MDL Number:MFCD00007526