Order Entry
France
ContactUsLinkComponent
Organic analytical standards, Paraben target analyte mix solution, reference material, Honeywell Fluka™
Organic analytical standards, Paraben target analyte mix solution, reference material, Honeywell Fluka™
Catalog # HONCR1069-1ML
CAS Number:  
Organic analytical standards, Paraben target analyte mix solution, reference material, Honeywell Fluka™
Catalog # HONCR1069-1ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Description:
    Paraben target analyte mix solution
  • Storage temperature:
    –20 °C
  • Description:
    Paraben target analyte mix solution
  • Flash point:
    –17 °C
  • Boiling point:
    55...57 °C
  • Freezing point:
    –95 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results