
Nicotinic acid (Niacin) 98.0-102.0% (dried basis), powder USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:59-67-6
:
Formula:C₆H₅NO₂
MDL Number:MFCD00006391
:123.11 g/mol
:GHS07
Specification Test Results
- Assay (Dried Basis):98.0 - 102.0 %
- Loss On Drying:max.1.0 %
- Residue On Ignition:max. 0.1 %
- Chloride (Cl):max. 0.02 %
- Sulfates (SO₄):max. 0.02 %
- Cadmium:As reported
- Lead (Pb):As reported
- ARSENIC (As):As reported
- Mercury (Hg):As reported
- Related compounds:To pass test
- Identification:To pass test
- Residual solvents:To pass test
Specification Test Results
Frequently Bought Together