5-Carboxy-X-rhodamine triethylammonium salt single isomer ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90006.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
- Environmentally Preferable:
Specifications
Specification Test Results
- Molecular Weight:635.6
Specification Test Results
About this item
Synonyms: 5-ROX triethylammonium salt, Carboxy-X-rhodamine triethylammonium salt, 5-isomer
:
:635.79 g/mol
Storage Temperature:Refrigerator
Formula:C₃₉H₄₅N₃O₅