Order Entry
France
ContactUsLinkComponent
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥95% (by TLC) DNA/RNA binding dye
Toxic
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥95% (by TLC) DNA/RNA binding dye
Catalog # 40041.
Supplier:  Biotium
CAS Number:  
undefined
Toxic
3,3'-Diethylthiadicarbocyanine iodide [DiSC₂(5)] ≥95% (by TLC) DNA/RNA binding dye
Catalog # 40041.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :518.48 g/mol
Storage Temperature:Refrigerator
EINECS:208-186-7
UN:2811
ADR:6.1,II
 :GHS06
CAS Number:514-73-8
Formula:C₂₃H₂₃N₂S₂I
MDL Number:MFCD00074829