Order Entry
France
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
Catalog # 40043.
Supplier:  Biotium
CAS Number:  
undefined
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 10 mg/mL in water used in nuclear staining
Catalog # 40043.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :350.25 g/mol
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681