4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) fluorescent dye, used in nuclear staining
Catalog # 40009.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:457.48 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·₂C₃H₆O₃