Order Entry
France
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) ≥98% (by HPLC) used in nuclear staining, Sigma-Aldrich®
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) ≥98% (by HPLC) used in nuclear staining, Sigma-Aldrich®
Catalog # D9564-10MG
CAS Number:  
undefined
4',6'-Diamidino-2-phenylindole dilactate (DAPI dilactate) ≥98% (by HPLC) used in nuclear staining, Sigma-Aldrich®
Catalog # D9564-10MG
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :457.48 g/mol
 :M-Clarity™ quality level = MQ300
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·₂C₃H₆O₃