Fura 2-AM ≥98% (by HPLC) fluorescent indicator, for calcium determination
Supplier: Biotium
Certificates
About this item
Synonyms: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester
CAS Number:108964-32-5
:
Formula:C₄₄H₄₇N₃O₂₄
MDL Number:MFCD00036976
Molecular Weight:1001.857 g/mol
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together