4-Bromo-A-23187 free acid ≥98% (by TLC, NMR) ionophore, for calcium determination
Catalog # 59006.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:602.52 g/mol
CAS Number:76455-48-6
Formula:C₂₉H₃₆BrN₃O₆