Supelco® TraceCERT® Organic Reference Standard, Nicotinic acid
79838-50MG
: Merck
:
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:59-67-6
- Description:Nicotinic acid
- Storage conditions:15...25 °C
- Hazard class description:H319
- Shipping conditions:STD
- MDL No.:MFCD00006391
- M-Clarity quality level:MQ300
- Synonym(s):3-Picolinic acid; Niacin; Pellagra preventive factor; Pyridine-3-carboxylic acid; Vitamin B₃
- Melting point:236 - 239 °C (lit.)
- Molecular weight:123,11
- Flash point:193 °C - Closed cup
- GHS signal: GHS07
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P305 + P351 + P338