Order Entry
France
ContactUsLinkComponent
Fluorescamine ≥95% (by TLC) amine-reactive fluorescent dye
Fluorescamine ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90092.
Supplier:  Biotium
CAS Number:  
Fluorescamine ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90092.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :278.26 g/mol
Storage Temperature:Freezer
CAS Number:38183-12-9
 :MFCD01860207
Formula:C₁₇H₁₀O₄
MDL Number:MFCD00005928