0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, prednisolone, (CRM), Supelco®
Pharmaceutical secondary standard, prednisolone, (CRM), Supelco®
PHR1043-500MG
:
Merck
:
Pharmaceutical secondary standard, prednisolone, (CRM), Supelco®
PHR1043-500MG
:
Merck
:
Pk:
500 mg
CAS No.:
50-24-8
Description:
Prednisolone
Description:
Prednisolone