0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Standards
Organic Standards
Pharmaceutical secondary standard, Bicalutamide, (CRM), Supelco®
Pharmaceutical secondary standard, Bicalutamide, (CRM), Supelco®
PHR1678-1G
:
Merck
:
Pharmaceutical secondary standard, Bicalutamide, (CRM), Supelco®
PHR1678-1G
:
Merck
:
Pk:
1 g
CAS No.:
90357-06-5
Description:
Bicalutamide
Description:
Bicalutamide