Supelco® TraceCERT® Organic Reference Standard, mono-Ethyl fumarate
Catalog # 92585-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:2459-05-4
- Description:mono-Ethyl fumarate
- Storage conditions:15...25 °C
- Hazard class description:H315 - H317 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00002699
- M-Clarity quality level:MQ300
- Synonym(s):Fumaric acid monoethyl ester; Monoethyl fumarate
- Melting point:66 - 68 °C (lit.)
- Formula:C₂H₅OCOCH=CHCOOH
- Molecular weight:144,13
- Boiling point:147 °C/16 mmHg (lit.)
Specifications
About this item
- GHS signal: GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P280 - P302 + P352 - P305 + P351 + P338