Supelco® Drug and metabolite standards, Sibutramine hydrochloride solution, (CRM)
S-011-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:125494-59-9
- Description:Sibutramine hydrochloride solution
- Description:Sibutramine hydrochloride solution
- Pk Info:ampoule of 1 ml