Supelco® Drug and metabolite standards, N-desmethyl-cis-tramadol hydrochloride solution, (CRM)
D-023-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1018989-94-0
- Description:N-Desmethyl-cis-tramadol hydrochloride solution
- Environmentally Preferable:
- Description:N-Desmethyl-cis-tramadol hydrochloride solution
- Pk Info:ampoule of 1 ml