Drug and metabolite standards, (+/-)-4-methylephedrine hydrochloride solutions, (CRM), Supelco®
Catalog # M-160-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1292841-87-2
- Description:(+/-)-4-Methylephedrine-D3 hydrochloride solution
- Environmentally Preferable:
- Description:(+/-)-4-Methylephedrine-D3 hydrochloride solution
- Pk Info:ampoule of 1 ml