Order Entry
France
ContactUsLinkComponent
 
Columns, HiScreen™ Capto PlasmidSelect
  29-2017-90
 :  Cytiva
undefined
Columns, HiScreen™ Capto PlasmidSelect
  29-2017-90
 :  Cytiva

 

  • Flow rate:
    <2,3 mL/min
  • Max. pressure:
    8 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto PlasmidSelect
  • Bed dimensions:
    7,7×100 mm
  • Pk:
    1

 

 

Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiScreen™ columns

Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.