Order Entry
France
ContactUsLinkComponent
Gram's iodine solution for microscopy, Sigma-Aldrich®
Gram's iodine solution for microscopy, Sigma-Aldrich®
Certificates
Gram's iodine solution for microscopy, Sigma-Aldrich®
Certificates

Specification Test Results

  • Appearance (colour):
    Brown to very dark brown
  • Appearance (form):
    Liquid
  • Density D20/4:
    1.003 - 1.007
  • Refractive index N20/D:
    1.334 - 1.336

Specification Test Results

Related Information

Frequently Bought Together