Sterilisation flat reels, Tyvek®/film, Stericlin®
Supplier: VP Medical Packaging
Certificates
About this item
These see-through sterilisation reels are made of a medical grade of Tyvek® non woven polyethylene fabric, Type 1073 B with 75 g/m².
- Effective barrier against germs, bacteria and moisture
- High resistance to penetration and fibre-free peeling
- Not suitable for steam sterilisation
This is fully compatible with the requirements of VH₂O₂ (plasma) sterilisation and irradiation. This sterile barrier system is the first choice for gas sterilisation, for Tyvek® hardly absorbs any toxic gases and thus results in residual toxic gas levels that are far below the legal limit values.
Certifications: ISO 11607. CE.
Frequently Bought Together