Order Entry
France
ContactUsLinkComponent
Size exclusion chromatography columns, HiLoad™ 16/600 Superose™ 6 Prep Grade (pg)
Size exclusion chromatography columns, HiLoad™ 16/600 Superose™ 6 Prep Grade (pg)
Catalog # 29-3239-52
Supplier:  Cytiva
undefined
Size exclusion chromatography columns, HiLoad™ 16/600 Superose™ 6 Prep Grade (pg)
Catalog # 29-3239-52
Supplier:  Cytiva

Specifications

  • Flow rate:
    1,6 ml/min
  • Max. pressure:
    3 bar
  • Capacity:
    120 ml
  • Description:
    HiLoad™ 16/600 Superose™ 6 prep grade
  • pH range:
    3 - 12
  • Storage:
    4 to 30 ˚C
  • Column hardware:
    Glass
  • Bed dimensions:
    16×600 mm
  • Flow velocity:
    <40 cm/h
  • Pk:
    1
  • Sample volume:
    <5 ml

Specifications

About this item

HiLoad™ 16/600 Superose™ 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 ml.

  • Convenience and reproducibility: can be used with standalone pump or a chromatography system
  • High-resolution separations: purification is possible across an exceptionally broad molecular-weight range
  • Scalable: separations can be scaled up using Superose™ 6 prep grade resin
  • ÄKTA system compatible: fittings (0.062") for direct connection to ÄKTA systems and other chromatography systems

This column is packed with Superose™ 6 pg resin, which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5000000.