Eyewash solution
Eyewash solution, 500 ml bottle
LBES8205261
Eyewash bottle containing 0,9% sodium chloride solution.
- Ergonomic eye-cups for effective flow
- Sealed to ensure the integrity of the device
- Easier to open the eye for optimal efficiency
Suitable for dust or body wash
Rinsing time: Five minutes
Shelf life: 3 years
- Description:Eyewash solution, 500 ml bottle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



