Order Entry
France
ContactUsLinkComponent
 
 
 
 
Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™
 
 
 
 
Piperidine ≥99.0% (by GC, titration analysis), Puriss. p.a., Riedel-de Haën™

 

  • Refractive Index:
    n20/D 1.452 (lit.)
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.452 - 1.454
  • Non-volatile matter:
    Max. 0.02 %
  • Water (Karl Fischer):
    Max. 0.3 %
  • Aluminium (Al):
    Max. 0.5 ppm
  • Barium (Ba):
    Max. 0.1 ppm
  • Bismuth (Bi):
    Max. 0.1 ppm
  • Calcium (Ca):
    Max. 0.5 ppm
  • Cadmium (Cd):
    Max. 0.05 ppm
  • Cobalt (Co):
    Max. 0.02 ppm
  • Chromium (Cr):
    Max. 0.02 ppm
  • Copper (Cu):
    Max. 0.02 ppm
  • Iron (Fe):
    Max. 0.1 ppm
  • Potassium (K):
    Max. 0.5 ppm
  • Lithium (Li):
    Max. 0.1 ppm
  • Magnesium (Mg):
    Max. 0.1 ppm
  • Manganese (Mn):
    Max. 0.02 ppm
  • Molybdenum (Mo):
    Max. 0.1 ppm
  • Sodium (Na):
    Max. 0.5 ppm
  • Nickel (Ni):
    Max. 0.02 ppm
  • Lead (Pb):
    Max. 0.1 ppm
  • Strontium (Sr):
    Max. 0.1 ppm
  • Tin (Sn):
    Max. 0.1 ppm

 

 

Frequently Bought Together