Order Entry
France
ContactUsLinkComponent
 
 
Ammonium chloride 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 
undefined
 
Ammonium chloride 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

 

  • Assay (calculated to the dried substance):
    99.5 - 100.5 %
  • Loss on drying (105°C):
    Max. 0.5 %
  • Loss on drying (on silicagel, 4h):
    Max. 0.5 %
  • Residue on ignition (SO₄):
    Max. 0.1 %
  • pH (5 %, 25 °C):
    4.6 - 6.0
  • Acidity or alkalinity:
    complying
  • Arsenic (As):
    Max. 0.0003 %
  • Calcium (Ca):
    Max. 100 ppm
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 20 ppm
  • Magnesium (Mg):
    Max. 0.001 %
  • Zinc (Zn):
    Max. 0.002 %
  • Lead (Pb):
    Max. 0.0004 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Iodide and bromide:
    complying
  • Sulfate (SO₄):
    Max. 100 ppm
  • Thiocyanate (SCN):
    complying
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

 

 

Frequently Bought Together