Order Entry
France
ContactUsLinkComponent
 
 
 
 
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
 
undefined
 
 
 
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
 

 

  • Vapor Pressure:
    26 mmHg ( 25 °C)
  • Assay (GC):
    Min. 99.7 %
  • Density (D 20/20):
    0.865 - 0.869
  • Refractive index (n 20/D):
    1.496 - 1.498
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as HCl):
    Max. 0.001 %
  • Free alkali (as NaOH):
    Max. 0.002 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Barium (Ba):
    Max. 0.00001 %
  • Bismuth (Bi):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Potassium (K):
    Max. 0.00005 %
  • Lithium (Li):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Molybdenum (Mo):
    Max. 0.00001 %
  • Sodium (Na):
    Max. 0.00005 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Strontium (Sr):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • S-compounds (as S):
    Max. 0.003 %
  • Thiophene:
    Max. 0.0001 %
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10

 

 

Frequently Bought Together