Order Entry
France
ContactUsLinkComponent
Chloroform ≥99.5% stabilised
Chloroform ≥99.5% stabilised
Certificates
undefined
Chloroform ≥99.5% stabilised
Certificates

Specification Test Results

  • Vapor Pressure:
    160 mmHg ( 20 °C)
  • NMR-spectrum:
    complying
  • Assay (GC):
    Min. 99.50 %
  • 2-methyl-2-buten (GC):
    20 - 200 ppm
  • Refractive Index:
    n20/D 1.445 (lit.)

Specification Test Results

Related Information

Frequently Bought Together