Order Entry
France
ContactUsLinkComponent
di-Sodium tetraborate decahydrate ≥99.5% ACS, Fluka™
di-Sodium tetraborate decahydrate ≥99.5% ACS, Fluka™
Certificates
di-Sodium tetraborate decahydrate ≥99.5% ACS, Fluka™

Specification Test Results

  • Assay:
    99.5 - 105.0 %
  • pH (0.01 m, 25°C):
    9.15 - 9.20
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 50 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 10 ppm
  • Phosphate (PO₄):
    Max. 10 ppm

Specification Test Results

Related Information

Frequently Bought Together