Analytical standard, Methyl 4-hydroxybenzoate-ring-¹³C₆ solution, reference material, Honeywell Fluka™
Catalog # HONCR1347-5ML
Fournisseur: Honeywell Chemicals
Numéro CAS:
Spécifications
- Pack type:Glass Bottle
- Pk:5 mL
- Standards:Analytical standard, single component solution
- CAS No.:1581694-95-2
- Description:Methyl 4-hydroxybenzoate-ring-¹³C₆ solution, 10 μg/ml in acetone
- Melting point:–95 °C
- Formula:¹³C₆C₂H₈O₃
- Storage temperature:2…8 °C
- Description:Methyl 4-hydroxybenzoate-ring-¹³C₆ solution
- Boiling point:55…57 °C
Spécifications
À propos de cet article
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results