Analytical standard, Diclofenac-(acetophenyl ring-¹³C₆) sodium salt hydrate, reference material, Honeywell Fluka™
HONCR1265-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1261393-73-0
- Description:Diclofenac-(acetophenyl ring-¹³C₆) sodium salt hydrate
- Melting point:min. 275 °C
- Formula:¹³C6C8H10Cl2NNaO2 · 4.5H2O
- Storage temperature:Ambient
- Description:Diclofenac-(acetophenyl ring-¹³C₆) sodium salt hydrate
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results