Chiral HPLC columns, NUCLEOCEL DELTA
Cellulose-modified silica phases for HPLC enantiomer separation.
- Base material: silica gel
- Chiral selector: cellulose tris-(3,5-dimethylphenylcarbamate)
- Phases for normal phase or reversed phase mode
NUCLEOCEL DELTA is recommended for enantiomer separation methods in reference to a successful application note and for chiral method developments of pharmaceutically active compounds, pollutants, food ingredients, catalysts, and bioorganic compounds.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
- Catalog No:
- MANA721186.30
- MANA720446.46
- MANA720450.46
- MANA720451.46
- MANA720445.46
- MANA721185.30
- USP code:
- L40
- L40
- L40
- L40
- L40
- L40
Frequently Bought Together