HPLC columns, NUCLEOSIL® SB
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEOSIL® SB is a quaternary ammonium modified silica phase (SAX) for HPLC.
- Robust spherical silica gel
- Anionic phase for ion exchange chromatography
- Separation mechanism based on strong ion exchange interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Capacity: ~1 meq/g
Specifications
- Catalog No:
- MANA721025.30
- MANA720996.46
- MANA720029.46
- MANA720989.46
- MANA720183.46
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- 10%
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L14
- L14
- L14
- L14
- L14