Order Entry
France
ContactUsLinkComponent
Poly(methyl methacrylate), M.W. 90000∼, fine granules secondary standard
Poly(methyl methacrylate), M.W. 90000∼, fine granules secondary standard
Catalog # ACRO178780250
CAS Number:  
undefined
Poly(methyl methacrylate), M.W. 90000∼, fine granules secondary standard
Catalog # ACRO178780250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White Beads
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 %
  • Particle size:
    50 to 150 microns (Approximately)

Specification Test Results

About this item

 :
MDL Number:MFCD00134349
Melting Pt:>150 °C
Flash Pt:303 °C
Storage Temperature:Ambient
CAS Number:9011-14-7
Formula:C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)