Streptococcus pyogenes Cas9 nuclease
About this item
Optimised Cas9 Nuclease S. Pyogenes is designed for genome editing in living cells or organisms and also for in vitro digestion.
- Increased genome editing efficiency using Cas9/RNP delivery
- Successful CRISPR/Cas9 genome editing can be performed through diverse approaches (plasmids, mRNA, nuclease, viral delivery)
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). These features make nuclease protein delivery particularly well suited for precision genome engineering.
Efficient nucleic acid delivery represents a critical step for genome editing experiments.
Informations de livraison: Shipped in dry ice.
Attention: Cas9 nuclease and Cas9 reaction buffer must be stored at –20 °C. Avoid freeze/thawing cycles.
3 Options Available Below
Product Details & Documents
Optimised Cas9 Nuclease S. Pyogenes is designed for genome editing in living cells or organisms and also for in vitro digestion.
- Increased genome editing efficiency using Cas9/RNP delivery
- Successful CRISPR/Cas9 genome editing can be performed through diverse approaches (plasmids, mRNA, nuclease, viral delivery)
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). These features make nuclease protein delivery particularly well suited for precision genome engineering.
Efficient nucleic acid delivery represents a critical step for genome editing experiments.
Informations de livraison: Shipped in dry ice.
Attention: Cas9 nuclease and Cas9 reaction buffer must be stored at –20 °C. Avoid freeze/thawing cycles.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- OZBICAS9050
- OZBICAS9100
- OZBICAS9500
- Enzyme name:
- Cas9 Nuclease
- Cas9 Nuclease
- Cas9 Nuclease
- Species:
- Streptococcus pyogenes
- Streptococcus pyogenes
- Streptococcus pyogenes
- Molecular weight:
- 164,48 kDa
- 164,48 kDa
- 164,48 kDa
- Form:
- Lyophilized powder
- Lyophilized powder
- Lyophilized powder
- Storage conditions:
- −20 °C
- −20 °C
- −20 °C
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.OZBICAS9050Enzyme nameCas9 NucleaseSpeciesStreptococcus pyogenesMolecular weight164,48 kDaFormLyophilized powderStorage conditions−20 °C - Specifications:
More
Specifications
Cat. No.OZBICAS9100Enzyme nameCas9 NucleaseSpeciesStreptococcus pyogenesMolecular weight164,48 kDaFormLyophilized powderStorage conditions−20 °C - Specifications:
More
Specifications
Cat. No.OZBICAS9500Enzyme nameCas9 NucleaseSpeciesStreptococcus pyogenesMolecular weight164,48 kDaFormLyophilized powderStorage conditions−20 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



