Order Entry
France
ContactUsLinkComponent
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Irritant
Corrosive
Environment
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # 4895-22
CAS Number:  
undefined
Irritant
Corrosive
Environment
Copper(II) bromide 50% in aqueous solution, AR®, Macron Fine Chemicals™
Catalog # 4895-22
CAS Number:  

Specifications

  • Pack type:
    HDPE drum
  • Pk:
    50 lb
  • Seq:
    7137

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    49 - 51%
  • Appearance (dark brown/black solution):
    Passes Test
  • Assay (CuBr₂):
    49 - 51 %
  • Insoluble Matter:
    ≤0.005 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Chloride (Cl):
    ≤0.15 %
  • Substances not Precipitated by H₂S (as SO₄):
    ≤0.10 %
  • Iron (Fe):
    ≤0.003 %
  • pH of 10% solution:
    2.7 - 3.0

Specification Test Results

About this item

 :
 :223.35 g/mol
Density:1.50 kg (1 L)
UN:3264
ADR:8,II
 :GHS07
 :GHS05
 :GHS09
CAS Number:7789-45-9
Formula:Br₂Cu
MDL Number:MFCD00010970