Order Entry
France
ContactUsLinkComponent
SPE columns, acrylamide analysis, CHROMABOND® ABC18
SPE columns, acrylamide analysis, CHROMABOND® ABC18
Catalog # MANA730533
Supplier:  MACHEREY-NAGEL
undefined
SPE columns, acrylamide analysis, CHROMABOND® ABC18
Catalog # MANA730533
Supplier:  MACHEREY-NAGEL
Specifications
  • Capacity:
    6 ml
  • Bed weight:
    500 mg
  • Column hardware:
    PP
  • Phase:
    ABC18
  • Pk:
    30
Specifications
About this item
CHROMABOND® ABC18 is a special phase for SPE analysis of acrylamide in food.
  • Special octadecyl phase
  • Additional ion exchange functions
Application: Clean-up of acrylamide from ultra-heated starch-containing food, such as potato chips, french fries, crispbread, and cereals.