Broxettes of evaporated salt tablets
Catalog # KAMM1161131
Supplier: KAMMERER
Specifications
- Description:Broxettes of evaporated salt tablets
- Environmentally Preferable:
- Packed:25 kg bag
- Pk:1
Specifications
About this item
Pressed evaporated salt tablets for the regeneration of ion exchangers.
- Highest degree of purity
- Tested and suitable for chlorine chlorinators (according to EN 973 type A)