SPE columns, cannabis analysis, CHROMABOND® Drug II
MANA730680
- Particle Size:45 µm
- Pore size:60 Å
- Capacity:3 ml
- Seq:02
- pH range:2 - 8
- Surface area:500 m²/g
- Bed weight:200 mg
- Column hardware:PP
- Phase:Drug II
- Pk:50
CHROMABOND® Drug II is a special silica phase for the SPE of THC and its derivatives from biological fluids.
- Base material: Silica
- Special bifunctional modification: C8 / SB (strong anion exchanger)
Application: Extraction of THC and its derivatives from urine, blood, serum, and plasma as well as acidic analytes from biological fluids.