Order Entry
France
ContactUsLinkComponent
 
 
 
Sodium silicate solution, Reagent Grade
 
 
 
Sodium silicate solution, Reagent Grade

 

  • Assay of NaOH:
    13.4 - 14.4 %
  • Assay of SiO₂:
    25.7 - 27.8 %
  • Composition:
    SiO₂, ~26.5%

 

Frequently Bought Together