Calcium Ionophore IV - Cocktail for membranes, Selectophore™ ion sensor material, Supelco®
Catalog # 70356-1ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Seq:2070
Specifications
Specification Test Results
- APPEARANCE (COLOR):Colorless to Yellow and Colorless to Green-Yellow
- APPEARANCE (FORM):Liquid
- APPLICATION TEST:CORRESPONDS TO REQUIREMENTS
Specification Test Results
About this item
:
Flash Pt:–18 °C
UN:2056
ADR:3,II
:GHS02
:GHS07
:GHS08