Order Entry
France
ContactUsLinkComponent
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Flammable
Health
Irritant
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Catalog # VWRS28552.410
Supplier:  VWR Collection
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Catalog # VWRS28552.410
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Boiling point:
    65.0 - 66.5 °C
  • Density (20/4):
    0.885 - 0.888
  • Free acidity:
    Max. 0.0003 meq/g
  • n 20/D:
    1.406 - 1.408
  • Peroxides (as H2O2):
    Max. 100 ppm
  • Water:
    Max. 0.03 %

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108,5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250-400 ppm
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356