Order Entry
France
ContactUsLinkComponent
Ethanol ≥70% (v/v), GPR RECTAPUR®
Flammable
Ethanol ≥70% (v/v), GPR RECTAPUR®
Catalog # VWRS98193.450
Supplier:  VWR Collection
CAS Number:  
Flammable
Ethanol ≥70% (v/v), GPR RECTAPUR®
Catalog # VWRS98193.450
Supplier:  VWR Collection
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Assay (V/V):
    69 - 71 %
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

About this item

 :
 :46.07 g/mol
Density:0.85 g/cm³ (20 °C)
Flash Pt:22 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568