Order Entry
France
ContactUsLinkComponent
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Catalog # VWRS25107.255
Supplier:  VWR Collection
CAS Number:  
Magnesium chloride hexahydrate ≥97%, GPR RECTAPUR®
Catalog # VWRS25107.255
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    545 g
  • Seq:
    3680

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Alkaline metals (as SO4):
    Max. 2.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Ca (Calcium):
    Max. 0.1 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781