Order Entry
France
ContactUsLinkComponent
Inlet tubing kit
Inlet tubing kit
  18-1119-26
 :  Cytiva
undefined
Inlet tubing kit
  18-1119-26
 :  Cytiva

 

  • Ext.Ø:
    4,76 mm
  • Ø int.:
    1,6 mm
  • Length:
    1,5 m
  • Pk:
    1

 

 

Made of Perfluoroethylene-propylene copolymer (FEP).

The tubing is fitted with: 2 x Nut UNF 5/16" male 2 x ferrule. The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran.