Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Wrenches
Wrench
Wrench
RIXI8-6018-5161
:
RIXIUS
Wrench
RIXI8-6018-5161
:
RIXIUS
Jaw size:
61 mm
Description:
Wrench
Material:
HDPE
Pk:
1
Wrench for 20 to 30 L canisters with 61 mm thread.