Order Entry
France
ContactUsLinkComponent
3-Amino-2-methylpyridine 97%
3-Amino-2-methylpyridine 97%
Catalog # H27540.03
CAS Number:  
3-Amino-2-methylpyridine 97%
Catalog # H27540.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :108.14 g/mol
Melting Pt:114…119 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:3430-10-2
Formula:C₆H₈N₂
MDL Number:MFCD03788195