2-Amino-5-methylpyridine
Catalog # MOLE36300968-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:108.14 g/mol
Boiling Pt:227.1 °C (1013 hPa)
Melting Pt:74…77 °C
Storage Temperature:Ambient
EINECS:216-503-5
UN:2811
ADR:6.1,III
CAS Number:1603-41-4
Formula:C₆H₈N₂
MDL Number:MFCD00006328