2-Amino-5-methylpyridine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1603-41-4
:
Formula:C₆H₈N₂
MDL Number:MFCD00006328
:108.14 g/mol
Boiling Pt:227.1 °C (1013 hPa)
Melting Pt:80…81 °C
Storage Temperature:Ambient
EINECS:216-503-5
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:72 to 77 °C
- Water:≤0.5 %
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together