2-Amino-3-methylpyridine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1603-40-3
:
Formula:C₆H₈N₂
MDL Number:MFCD00006320
:108.14 g/mol
Boiling Pt:221.1 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1,073 g/cm³ (20 °C)
Flash Pt:111 °C
Storage Temperature:Ambient
EINECS:216-501-4
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Clear yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.5813 to 1.5833 (20°C, 589 nm)
- Water:≤0.8 %
- GC:≥95 %
- Fluoride (F):≤10 ppm
- Arsenic (As):≤3 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤5 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.3 % (in water)
Specification Test Results
Frequently Bought Together